4-Thiazolecarboxylic acid, 97%, Thermo Scientific Chemicals
4-Thiazolecarboxylic acid, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

4-Thiazolecarboxylic acid, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
Catalog number 443970050
Price (USD)/ Each
247.00
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
247.00
Add to cart
4-Thiazolecarboxylic acid, 97%, Thermo Scientific Chemicals
Catalog number443970050
Price (USD)/ Each
247.00
-
Add to cart
Chemical Identifiers
CAS3973-08-8
IUPAC Name1,3-thiazole-4-carboxylic acid
Molecular FormulaC4H3NO2S
InChI KeyHMVYYTRDXNKRBQ-UHFFFAOYSA-N
SMILESOC(=O)C1=CSC=N1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to yellow to beige
Melting point190°C to 205°C
Loss on drying=<3 %
Infrared spectrumConforms
HPLC>=96.0 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only