Thermo Scientific Chemicals

2,4,5-Trichloropyrimidine, 98%, Thermo Scientific Chemicals

Catalog number: 443990250
25 g, Each
Thermo Scientific Chemicals

2,4,5-Trichloropyrimidine, 98%, Thermo Scientific Chemicals

Catalog number: 443990250
25 g, Each
Quantity
Catalog number: 443990250
Price (USD)
Have Questions?

Chemical Identifiers

CAS
5750-76-5
IUPAC Name
2,4,5-trichloropyrimidine
Molecular Formula
C4HCl3N2
InChI Key
GIKMWFAAEIACRF-UHFFFAOYSA-N
SMILES
ClC1=NC=C(Cl)C(Cl)=N1
Appearance (Form)
Liquid
GC
>=97.5 %
Infrared spectrum
Conforms
Appearance (Color)
Colorless

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text