Applied Biosystems™

3500 Genetic Analyzer for Sequence Typing & Fragment Analysis

Catalog number: 4440470
Have Questions?
Applied Biosystems™

3500 Genetic Analyzer for Sequence Typing & Fragment Analysis

Catalog number: 4440470
Have Questions?
Catalog Number
4440470
Unit Size
1 system
Price (USD)
Catalog NumberUnit SizePrice (USD)
44404701 systemRequest A Quote
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
This 8-capillary Genetic Analyzer is specifically configured with software for both fragment analysis and sequence typing applications.

We set a new standard in capillary electrophoresis with the 8-capillary 3500 Genetic Analyzer. These instruments are specifically designed to support the demanding performance needs of validated and process controlled environments, while retaining the unsurpassed application versatility that life science researchers expect from us.

• An 8 capillary system that can easily be upgraded to a 24-capillary system when you’re ready.
• New single-line 505 nm, solid-state long-life laser – utilizes a standard power supply; requires no heat-removal ducting.
• Powerful, integrated data collection and primary analysis software provides real-time assessment of data quality.
• Radio Frequency Identification (RFID) technology tracks key consumables data and records administrative information.
• Advanced multiplexing capabilities for DNA fragment analysis with up to six unique dyes.
• Unrivaled application flexibility – one array and one polymer are used for most applications.
• Simple setup, operation and maintenance – the easiest-to-run, easiest-to-own DNA sequencer to date.

Accurate Reliable Data Quality
The 3500 series systems significantly improves signal uniformity from instrument-to-instrument, run-to-run, and capillary-to-capillary. With a combination of intelligent hardware, powerful new algorithm and reagent combinations, these methods provide major reduction in the range of signal peak heights obtained across multiple 3500 series instruments (see figure 1).

Improved Sizing Precision
New thermal sub-system design gives enhanced performance in demanding fragment applications enabling improved sizing precision versus previous generations of capillary electrophoresis platforms (see figure 2).

Ready to Use Consumables With RFID Tags
The 3500 series introduces ready-to-use consumables with pre-packaged polymer pouches, cathode and anode buffer containers and easy-to-install capillary arrays. Each of these consumables is integrated with RFID tags that enable viewing, tracking and reporting of critical information about reagents and consumables including usage, lot number, part number, expiry date and on-instrument lifetime within the 3500 Series Data Collection Software (see figures 3 and 4).

Easy to Use New Data Collection Software
3500 Series Data Collection software breaks new ground in user-friendly navigation with an intuitive dashboard design, highly visible buttons for common operations, easy-to-read graphical displays to monitor the state of consumables and handy maintenance scheduling calendar functionality. New functionality includes simplified plate set-up, built-in primary analysis with quality control, so you can make decisions about the quality of data as it is produced on the instrument without the need to transfer output files into secondary analysis software packages (see figures 4 & 5).

For Research Use Only. Not for use in diagnostics procedures.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Software Type
Data Collection Software, Fragment Analysis
Starting Material
DNA
Amperage
15 A
Capillary Length
50 cm, 36 cm
Electrical Requirements
100 V to 240 V, 50/60 HZ, 15 A
External Computer
Hard Drive: 2x 500GB SATA 3.0 Gb/s and 8 MB Data Burst Cache, Memory: 16 GB (2x 8GB) 1600 MHz DDR3 Non-ECC. 4th Gen Intel Core I7 Processor- 3.1 GHz Turbo Processor, Operating System: Windows 7 SP1
Format
96-well Plate, 8-tube Strip
For Use With (Equipment)
3500 Genetic Analyzer Capillary Array (50 cm), POP-6™ Polymer for 3500⁄3500xL Genetic Analyzers, POP-4™ Polymer for 3500⁄3500xL Genetic Analyzers, 3500 Genetic Analyzer Capillary Array (36 cm), POP-7™ Polymer for 3500⁄3500xL Genetic Analyzers
Height (English) Exterior
28.34 in.
Height (Metric) Exterior
72 cm
Length (English) Exterior
24.01 in.
Length (Metric) Exterior
61 cm
Max. Voltage
Electrophoresis Voltage - Up to 20 kV
Number of Capillaries
8 Capillaries
Operating Environment
Humidity: 20%-80% (non-condensing), Room temp: <2° fluctuation during runs, Temperature: 15°C-30°C
Operating System
Windows 7 (Service Pack 1)
Polymer
POP-4™, POP-6™, POP-7™
Polymer Consumption
Replace at earliest of 7 days⁄384 samples⁄60 Injections (384-sample pouches), Replace at earliest of 7 days⁄960 samples⁄120 Injections (960-sample pouches)
Quantity
1 System
Read Length
≥850 bp using a 50 cm capillary and POP-7™
Software Compatibility
Sequencing Analysis Software, Variant Reporter™ Software, GeneMapper™ Software
Throughput
Up to 424 samples per day (using run module HID36 POP-7)
Warranty
1 Year Limited Warranty on Parts and Labor, Service Installation and Application Training Included
Weight
82 kg (approximately)
Width (English) Exterior
24.01 in.
Width (Metric) Exterior
61 cm

Package Components

4361142
Catalog Number:
4361142
Unit Size:
Qty:
1
Catalog Number:
4405673
Unit Size:
1 system
Qty:
1
Catalog Number:
4405778
Unit Size:
Qty:
1

Contents & Storage

This package consists of the 8 capillary instrument, the 3500 Data Collection Software, a Dell Workstation and monitor as well as reagent kits for system qualification. This package also includes GeneMapper™ software for fragment analysis, and software for sequence typing applications.

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text