Thermo Scientific Chemicals

3-(N-BOC-aminomethyl)-5-bromopyridine, 97%, Thermo Scientific Chemicals

Catalog number: 444060010
1 g, Each
Thermo Scientific Chemicals

3-(N-BOC-aminomethyl)-5-bromopyridine, 97%, Thermo Scientific Chemicals

Catalog number: 444060010
1 g, Each
Quantity
Catalog number: 444060010
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
943722-24-5
IUPAC Name
tert-butyl N-[(5-bromopyridin-3-yl)methyl]carbamate
Molecular Formula
C11H15BrN2O2
InChI Key
VZPOHASZICQBDX-UHFFFAOYSA-N
SMILES
CC(C)(C)OC(=O)NCC1=CC(Br)=CN=C1
Appearance (Color)
White to yellow
Infrared spectrum
Conforms
Appearance (Form)
Powder
GC
>=96.0 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text