This is highly deionized formamide used to resuspend samples before electrokinetic injection on capillary electrophoresis systems.For Research Use Only. NotRead more
Have Questions?
Change view
Catalog Number
Quantity
4440753
4 x 5 mL
4311320
25 mL
2 Options
Catalog number 4440753
Price (USD)
101.65
Online Exclusive
107.00
Save 5.35 (5%)
Each
Add to cart
Quantity:
4 x 5 mL
Price (USD)
101.65
Online Exclusive
107.00
Save 5.35 (5%)
Each
Add to cart
This is highly deionized formamide used to resuspend samples before electrokinetic injection on capillary electrophoresis systems.
For Research Use Only. Not for use in diagnostics procedures.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
DescriptionFormamide, 4x 5mL, Liquid, Formamide Reagent Type