1-Methyl-1H-pyrazole-4-carboxylic acid, 97%, Thermo Scientific Chemicals
1-Methyl-1H-pyrazole-4-carboxylic acid, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

1-Methyl-1H-pyrazole-4-carboxylic acid, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number 444080010
Price (USD)/ Each
-
Quantity:
1 g
Request bulk or custom format
Chemical Identifiers
IUPAC Name1-methyl-1H-pyrazole-4-carboxylate
Molecular FormulaC5H5N2O2
InChI KeyUPPPWUOZCSMDTR-UHFFFAOYSA-M
SMILESCN1C=C(C=N1)C([O-])=O
Molecular Weight (g/mol)125.11
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to light yellow
Appearance (Form)Powder
Infrared spectrumConforms
HPLC>=96.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only