Thermo Scientific Chemicals

Gallium(III) chloride, 99.99+%, (trace metal basis), Anhydrous, Beads, -10 mesh, Thermo Scientific Chemicals

Catalog number: 444101000
100 g, Glass bottle, Each
Thermo Scientific Chemicals

Gallium(III) chloride, 99.99+%, (trace metal basis), Anhydrous, Beads, -10 mesh, Thermo Scientific Chemicals

Catalog number: 444101000
100 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 444101000
Price (USD)
Have Questions?

Chemical Identifiers

CAS
13450-90-3
IUPAC Name
gallium(3+) trichloride
Molecular Formula
Cl3Ga
InChI Key
UPWPDUACHOATKO-UHFFFAOYSA-K
SMILES
[Cl-].[Cl-].[Cl-].[Ga+3]
Appearance (Color)
White to off-white
Appearance (Form)
Beads
Total trace metal impurities
=<100 ppm

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For ultra trace analysis
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text