Xylenes, technical
Xylenes, technical
Xylenes, technical
Xylenes, technical
Thermo Scientific Chemicals

Xylenes, technical

CAS: 1330-20-7 | C8H10 | 106.17 g/mol
Have Questions?
Catalog NumberQuantityPackaging
4442400505 LPlastic Drum
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 444240050
Price (USD)
-
Quantity:
5 L
Packaging:
Plastic Drum
Request bulk or custom format
Chemical Identifiers
CAS1330-20-7
IUPAC Name1,3-xylene
Molecular FormulaC8H10
InChI KeyIVSZLXZYQVIEFR-UHFFFAOYSA-N
SMILESCC1=CC(C)=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Density0.860 to 0.875 kg/l
GC>=98.5 % (Mixture of isomers)
Appearance (Form)Liquid
Appearance (Color)Clear colorless
Ask our AI about this Product
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link

1x1 image pixel for data collection