Thermo Scientific Chemicals

Water, technical, demineralized, Thermo Scientific Chemicals

Catalog number: 444300250
25 L, Plastic Can, Each
Thermo Scientific Chemicals

Water, technical, demineralized, Thermo Scientific Chemicals

Catalog number: 444300250
25 L, Plastic Can, Each
Quantity
Packaging
Catalog number: 444300250
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
7732-18-5
IUPAC Name
water
Molecular Formula
H2O
InChI Key
XLYOFNOQVPJJNP-UHFFFAOYSA-N
SMILES
O
pH
6.0 to 8.0
Appearance (Form)
Liquid
Conductivity
=<15 µS/cm
Appearance (Color)
Clear colorless

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text