Methanol, technical
Methanol, technical
Methanol, technical
Thermo Scientific Chemicals

Methanol, technical

CAS: 67-56-1 | CH4O | 32.04 g/mol
Have Questions?
Catalog NumberQuantityPackaging
4443100505 LPlastic Drum
Catalog number 444310050
Price (USD)
40.65
Special offer
Online exclusive
47.50
Save 6.85 (14%)
Each
Add to cart
Quantity:
5 L
Packaging:
Plastic Drum
Request bulk or custom format
Price (USD)
40.65
Special offer
Online exclusive
47.50
Save 6.85 (14%)
Each
Add to cart
Chemical Identifiers
CAS67-56-1
IUPAC Namemethanol
Molecular FormulaCH4O
InChI KeyOKKJLVBELUTLKV-UHFFFAOYSA-N
SMILESCO
View more
SpecificationsSpecification SheetSpecification Sheet
Refractive index1.3280 to 1.3300 (20°C, 589 nm)
Appearance (Form)Liquid
Water=<0.5 %
Appearance (Color)Clear colorless
GC>=99 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
RUO – Research Use Only

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link