Petroleum ether, technical, boiling range 100-140°C, Thermo Scientific Chemicals
Petroleum ether, technical, boiling range 100-140°C, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Petroleum ether, technical, boiling range 100-140°C, Thermo Scientific Chemicals

 
Have Questions?
Quantity:
25 L
Packaging:
Metal drum
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 444360250
Price (USD)/ Each
-
Quantity:
25 L
Packaging:
Metal drum
Request bulk or custom format
Chemical Identifiers
CAS64742-49-0, 111-65-9, 142-82-5, 108-87-2, 110-82-7, 73513-42-5
SpecificationsSpecification SheetSpecification Sheet
Density0.737 to 0.747 kg/l
Appearance (Color)Clear colorless
Boiling point98°C to 140°C (760 mmHg)
Appearance (Form)Liquid
Refractive index1.4070 to 1.4150 (20°C, 589 nm)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only