Citric acid monohydrate, technical, crystalline
Citric acid monohydrate, technical, crystalline
Citric acid monohydrate, technical, crystalline
Citric acid monohydrate, technical, crystalline
Thermo Scientific Chemicals

Citric acid monohydrate, technical, crystalline

CAS: 5949-29-1 | C6H8O7·H2O | 210.15 g/mol
Have Questions?
Catalog NumberQuantityPackaging
4443700505 kgPlastic Drum
Catalog number 444370050
Price (USD)
111.65
Online Exclusive
124.00
Save 12.35 (10%)
Each
In stock
Add to cart
Quantity:
5 kg
Packaging:
Plastic Drum
Request bulk or custom format
Price (USD)
111.65
Online Exclusive
124.00
Save 12.35 (10%)
Each
Add to cart
Chemical Identifiers
CAS5949-29-1
IUPAC Name2-hydroxypropane-1,2,3-tricarboxylic acid hydrate
Molecular FormulaC6H10O8
InChI KeyYASYEJJMZJALEJ-UHFFFAOYSA-N
SMILESO.OC(=O)CC(O)(CC(O)=O)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder or crystals or granules
Ask our AI about this Product
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Documents & Downloads

Certificates

Lot #Certificate TypeDateCatalog Number(s)
A0470240Certificate of AnalysisMay 17, 2025444370050
A0470417Certificate of AnalysisMay 17, 2025444370050
A0462103Certificate of AnalysisJun 17, 2024444370050
A0430564Certificate of AnalysisNov 27, 2023444370050
A0453981Certificate of AnalysisAug 25, 2023444370050
5 results displayed, search above for a specific certificate

Safety Data Sheets

Share catalog number, name or link

1x1 image pixel for data collection