Thermo Scientific Chemicals

4-Acetylphenylboronic acid pinacol ester, 97%, Thermo Scientific Chemicals

Catalog number: 444430010
1 g, Each
Thermo Scientific Chemicals

4-Acetylphenylboronic acid pinacol ester, 97%, Thermo Scientific Chemicals

Catalog number: 444430010
1 g, Each
Quantity
Catalog number: 444430010
Price (USD)
Have Questions?

Chemical Identifiers

CAS
171364-81-1
IUPAC Name
1-[4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]ethan-1-one
Molecular Formula
C14H19BO3
InChI Key
BATKIZWNRQGSKE-UHFFFAOYSA-N
SMILES
CC(=O)C1=CC=C(C=C1)B1OC(C)(C)C(C)(C)O1
Appearance (Form)
Solid
GC
>=96.0 %
NMR (1H-NMR)
Consistent with structure
Infrared spectrum
Conforms
Appearance (Color)
White to off-white

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text