Megaplex™ PreAmp Primers, Rodent Pool B v3.0
Product Image
Applied Biosystems™

Megaplex™ PreAmp Primers, Rodent Pool B v3.0

Green features
For use when assay sensitivity is of the utmost importance, Megaplex™ PreAmp Primers significantly enhance the ability to detect lowRead more
Have Questions?
Catalog number 4444308
Price (USD)/ 50 reactions
1,140.00
-
Add to cart
Price (USD)/ 50 reactions
1,140.00
Add to cart
Megaplex™ PreAmp Primers, Rodent Pool B v3.0
Catalog number4444308
Price (USD)/ 50 reactions
1,140.00
-
Add to cart
For use when assay sensitivity is of the utmost importance, Megaplex™ PreAmp Primers significantly enhance the ability to detect low expressed microRNAs enabling the generation of a comprehensive expression profile using far less sample, as low as 1 ng of input total RNA. With content matched to the Megaplex™ RT Primers, Megaplex™ PreAmp Primers, Rodent Pool B v3.0 is designed to be used with Megaplex™ RT Primers, Rodent Pool B v3.0. Following reverse transcription using Megaplex™ RT Primers, Megaplex™ PreAmp Primers along with the TaqMan® PreAmp Master Mix uniformly amplify all microRNAs prior to quantitation by real-time PCR using TaqMan® MicroRNA Arrays.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
GC-Rich PCR PerformanceLow
PCR MethodqPCR
Green FeaturesSustainable packaging
PrimerPreAmp Primers
Product LineMegaplex™, TaqMan™
Product TypePreamp Primer
Shipping ConditionRoom Temperature
SpeciesRat, Mouse
Sufficient For50 Reactions
No. of Reactions50 Reactions
Unit Size50 reactions
Contents & Storage
Store at -20°C