Thermo Scientific Chemicals

1-Acetyl-1H-pyrazole-4-boronic acid pinacol ester, 97%, Thermo Scientific Chemicals

Catalog number: 444440010
1 g, Each
Thermo Scientific Chemicals

1-Acetyl-1H-pyrazole-4-boronic acid pinacol ester, 97%, Thermo Scientific Chemicals

Catalog number: 444440010
1 g, Each
Quantity
Catalog number: 444440010
Price (USD)
Have Questions?

Chemical Identifiers

CAS
1150561-76-4
IUPAC Name
1-[4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazol-1-yl]ethan-1-one
Molecular Formula
C11H17BN2O3
InChI Key
GAXOQGDPSCOOLC-UHFFFAOYSA-N
SMILES
CC(=O)N1C=C(C=N1)B1OC(C)(C)C(C)(C)O1
NMR (1H-NMR)
Consistent with structure
Appearance (Form)
Solid
Appearance (Color)
White to light yellow
Infrared spectrum
Conforms
GC
>=96.0 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text