Thermo Scientific Chemicals

4-Amino-4'-chlorobiphenyl, 97%, Thermo Scientific Chemicals

Catalog number: 444460010
1 g, Each
Thermo Scientific Chemicals

4-Amino-4'-chlorobiphenyl, 97%, Thermo Scientific Chemicals

Catalog number: 444460010
1 g, Each
Quantity
Catalog number: 444460010
Price (USD)
Have Questions?

Chemical Identifiers

CAS
135-68-2
IUPAC Name
4'-chloro-[1,1'-biphenyl]-4-amine
Molecular Formula
C12H10ClN
InChI Key
OREQWMWYRYXCDF-UHFFFAOYSA-N
SMILES
NC1=CC=C(C=C1)C1=CC=C(Cl)C=C1
Appearance (Form)
Solid
NMR (1H-NMR)
Consistent with structure
Appearance (Color)
Off-white to orange to brown
Melting point
126°C to 132°C
GC
>=96 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text