2-Amino-5-chlorophenylboronic acid pinacol ester, 97%, Thermo Scientific Chemicals
2-Amino-5-chlorophenylboronic acid pinacol ester, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2-Amino-5-chlorophenylboronic acid pinacol ester, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number 444470010
Price (USD)/ Each
-
Quantity:
1 g
Request bulk or custom format
Chemical Identifiers
CAS1073371-77-3
IUPAC Name4-chloro-2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)aniline
Molecular FormulaC12H17BClNO2
InChI KeyGVJZHGCVSYBPIM-UHFFFAOYSA-N
SMILESCC1(C)OB(OC1(C)C)C1=C(N)C=CC(Cl)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to pink to tan
Infrared spectrumConforms
GC>=96.0 %
Appearance (Form)Solid
Melting point89°C to 95°C
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only