Thermo Scientific Chemicals

4-Amino-2-chlorophenylboronic acid pinacol ester, 97%, Thermo Scientific Chemicals

Catalog number: 444480050
5 g, Each
Thermo Scientific Chemicals

4-Amino-2-chlorophenylboronic acid pinacol ester, 97%, Thermo Scientific Chemicals

Catalog number: 444480050
5 g, Each
Quantity
Catalog number: 444480050
Price (USD)
Have Questions?

Chemical Identifiers

CAS
877160-63-9
IUPAC Name
3-chloro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)aniline
Molecular Formula
C12H17BClNO2
InChI Key
YKQXROMICPMQBZ-UHFFFAOYSA-N
SMILES
CC1(C)OB(OC1(C)C)C1=C(Cl)C=C(N)C=C1
Melting point
120°C to 125°C
Appearance (Form)
Crystalline powder
Infrared spectrum
Conforms
GC
>=96.0 %
Appearance (Color)
Off-white to light brown

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text