Thermo Scientific Chemicals

4-Amino-2-fluorophenylboronic acid pinacol ester, 97%, Thermo Scientific Chemicals

Catalog number: 444490010
1 g, Each
Thermo Scientific Chemicals

4-Amino-2-fluorophenylboronic acid pinacol ester, 97%, Thermo Scientific Chemicals

Catalog number: 444490010
1 g, Each
Quantity
Catalog number: 444490010
Price (USD)
Have Questions?

Chemical Identifiers

CAS
819057-45-9
IUPAC Name
3-fluoro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)aniline
Molecular Formula
C12H17BFNO2
InChI Key
FLMNWVXAEGUVNY-UHFFFAOYSA-N
SMILES
CC1(C)OB(OC1(C)C)C1=C(F)C=C(N)C=C1
Melting point
161°C to 167°C
NMR (1H-NMR)
Consistent with structure
Appearance (Color)
Off-white to brown
GC
>=96.0 %
Infrared spectrum
Conforms

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text