4-Amino-3-fluorophenylboronic acid pinacol ester, 97%, Thermo Scientific Chemicals
4-Amino-3-fluorophenylboronic acid pinacol ester, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

4-Amino-3-fluorophenylboronic acid pinacol ester, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number 444500010
Price (USD)/ Each
215.65
Special Offer
Online exclusive
Ends: 30-Jun-2025
254.00 
Save 38.35 (15%)
-
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (USD)/ Each
215.65
Special Offer
Online exclusive
Ends: 30-Jun-2025
254.00 
Save 38.35 (15%)
Add to cart
4-Amino-3-fluorophenylboronic acid pinacol ester, 97%, Thermo Scientific Chemicals
Catalog number444500010
Price (USD)/ Each
215.65
Special Offer
Online exclusive
Ends: 30-Jun-2025
254.00 
Save 38.35 (15%)
-
Add to cart
Chemical Identifiers
IUPAC Name2-fluoro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)aniline
Molecular FormulaC12H17BFNO2
InChI KeyAIXGNRNTXUKZLC-UHFFFAOYSA-N
SMILESCC1(C)OB(OC1(C)C)C1=CC(F)=C(N)C=C1
Molecular Weight (g/mol)237.08
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Solid
NMR (1H-NMR)Consistent with structure
Appearance (Color)Off-white to tan
Infrared spectrumConforms
GC>=96.0 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only