Thermo Scientific Chemicals

3-Amino-4-iodo-1H-pyrazole, 97%, Thermo Scientific Chemicals

Catalog number: 444520050
5 g, Each
Thermo Scientific Chemicals

3-Amino-4-iodo-1H-pyrazole, 97%, Thermo Scientific Chemicals

Catalog number: 444520050
5 g, Each
Quantity
Catalog number: 444520050
Price (USD)
Have Questions?

Chemical Identifiers

CAS
81542-51-0
IUPAC Name
4-iodo-1H-pyrazol-5-amine
Molecular Formula
C3H4IN3
InChI Key
CPVCVVMJPIOKMN-UHFFFAOYSA-N
SMILES
NC1=C(I)C=NN1
Infrared spectrum
Conforms
NMR (1H-NMR)
Consistent with structure
Appearance (Color)
Off-white to brown
Melting point
140°C to 146°C
Appearance (Form)
Solid

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text