3-Amino-2-methylphenylboronic acid pinacol ester, 97%, Thermo Scientific Chemicals
3-Amino-2-methylphenylboronic acid pinacol ester, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

3-Amino-2-methylphenylboronic acid pinacol ester, 97%, Thermo Scientific Chemicals

Have Questions?
Quantity:
5 g
Catalog number 444540050
Price (USD)/ Each
-
Quantity:
5 g
Request bulk or custom format
Chemical Identifiers
IUPAC Name2-methyl-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)aniline
Molecular FormulaC13H20BNO2
InChI KeyJMKMGPGFYMANCA-UHFFFAOYSA-N
SMILESCC1=C(C=CC=C1N)B1OC(C)(C)C(C)(C)O1
Molecular Weight (g/mol)233.12
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Solid
Infrared spectrumConforms
Melting point117°C to 122°C
NMR (1H-NMR)Consistent with structure
GC>=96.0 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only