Thermo Scientific Chemicals

4-Amino-3-nitrophenylboronic acid pinacol ester, 97%, Thermo Scientific Chemicals

Catalog number: 444570050
5 g, Each
Thermo Scientific Chemicals

4-Amino-3-nitrophenylboronic acid pinacol ester, 97%, Thermo Scientific Chemicals

Catalog number: 444570050
5 g, Each
Quantity
Catalog number: 444570050
Price (USD)
Have Questions?

Chemical Identifiers

CAS
833486-94-5
IUPAC Name
2-nitro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)aniline
Molecular Formula
C12H17BN2O4
InChI Key
QOYJKGGBFKVKDP-UHFFFAOYSA-N
SMILES
CC1(C)OB(OC1(C)C)C1=CC=C(N)C(=C1)[N+]([O-])=O
Appearance (Form)
Solid
Appearance (Color)
Yellow to orange to brown
Melting point
174°C to 180°C
GC
>=96.0 %
Infrared spectrum
Conforms

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text