Thermo Scientific Chemicals

4-[(tert-Butoxycarbonylamino)methyl]phenylboronic acid pinacol ester, 97%, Thermo Scientific Chemicals

Catalog number: 444640010
1 g, Each
Thermo Scientific Chemicals

4-[(tert-Butoxycarbonylamino)methyl]phenylboronic acid pinacol ester, 97%, Thermo Scientific Chemicals

Catalog number: 444640010
1 g, Each
Quantity
Catalog number: 444640010
Price (USD)
Have Questions?

Chemical Identifiers

CAS
330794-35-9
IUPAC Name
tert-butyl N-{[4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]methyl}carbamate
Molecular Formula
C18H28BNO4
InChI Key
CUDCEJRRWNIPDL-UHFFFAOYSA-N
SMILES
CC(C)(C)OC(=O)NCC1=CC=C(C=C1)B1OC(C)(C)C(C)(C)O1
Appearance (Color)
Colorless to white to off-white
Appearance (Form)
Solid
Infrared spectrum
Conforms
Melting point
66°C to 71°C
GC
>=96.0 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text