Thermo Scientific Chemicals

6-(tert-Butoxycarbonylamino)pyridine-3-boronic acid pinacol ester, 97%, Thermo Scientific Chemicals

Catalog number: 444660010
1 g, Each
Thermo Scientific Chemicals

6-(tert-Butoxycarbonylamino)pyridine-3-boronic acid pinacol ester, 97%, Thermo Scientific Chemicals

Catalog number: 444660010
1 g, Each
Quantity
Catalog number: 444660010
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
910462-31-6
IUPAC Name
tert-butyl N-[5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridin-2-yl]carbamate
Molecular Formula
C16H25BN2O4
InChI Key
USZVCDRKIVKICD-UHFFFAOYSA-N
SMILES
CC(C)(C)OC(=O)NC1=NC=C(C=C1)B1OC(C)(C)C(C)(C)O1
Appearance (Color)
White to off-white
Appearance (Form)
Solid
Infrared spectrum
Conforms
Melting point
202°C to 208°C
HPLC
>=96.0 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text