4-Cyano-2-fluorophenylboronic acid pinacol ester, 97%, Thermo Scientific Chemicals
4-Cyano-2-fluorophenylboronic acid pinacol ester, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

4-Cyano-2-fluorophenylboronic acid pinacol ester, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
Catalog number 444720050
Price (USD)/ Each
-
Quantity:
5 g
Request bulk or custom format
Chemical Identifiers
CAS1035235-29-0
IUPAC Name3-fluoro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzonitrile
Molecular FormulaC13H15BFNO2
InChI KeyFEUINZSEHIJUBU-UHFFFAOYSA-N
SMILESCC1(C)OB(OC1(C)C)C1=C(F)C=C(C=C1)C#N
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Colorless to white to yellow
Infrared spectrumConforms
Melting point82°C to 87°C
GC>=96.0 %
NMR (1H-NMR)Consistent with structure
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only