6-(4-Fluorophenyl)-2-pyridinecarboxaldehyde, 97%, Thermo Scientific Chemicals
6-(4-Fluorophenyl)-2-pyridinecarboxaldehyde, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

6-(4-Fluorophenyl)-2-pyridinecarboxaldehyde, 97%, Thermo Scientific Chemicals

Have Questions?
Catalog NumberQuantity
4447700101 g
Catalog number 444770010
Price (USD)
-
Quantity:
1 g
Request bulk or custom format
Chemical Identifiers
CAS834884-77-4
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Off-white to beige
Appearance (Form)Solid
Infrared spectrumConforms
Melting point86°C to 91°C
GC>=96.0 %
View more
Ask our AI about this Product
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link

1x1 image pixel for data collection