Applied Biosystems™

TaqMan™ Gene Expression Assay, VIC primer-limited

Catalog number: 4448486
Have Questions?
Applied Biosystems™

TaqMan™ Gene Expression Assay, VIC primer-limited

Catalog number: 4448486
Have Questions?
Catalog Number
4448486
Unit Size
L (2900 reactions), made to order
Price (USD)
Catalog Number
Unit Size
S (360 reactions), made to order
Price (USD)
Catalog Number
Unit Size
M (750 reactions), made to order
Price (USD)
Catalog NumberUnit SizePrice (USD)
4448486L (2900 reactions), made to order
4448484S (360 reactions), made to order
4448485M (750 reactions), made to order
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations

Applied Biosystems TaqMan Gene Expression Assays, VIC primer-limited, are used for quantitative real-time PCR analysis of gene expression and consist of a pair of unlabeled PCR primers and a TaqMan probe with a dye label (VIC) on the 5’ end and a minor groove binder (MGB) and nonfluorescent quencher (NFQ) on the 3’ end. These assays are primer-limited and ideal for multiplex reactions looking at two different gene targets in the same qPCR well.

Features include:
Easy to use—just add cDNA and master mix and run the qPCR—no melt curves required
Specific—TaqMan assays use proprietary MGB-containing probes that can be up to 15 bases shorter than non-MGB probes, improving the specificity of the assay
Sensitive—TaqMan assays are ideal for measuring low levels of expression or low-abundance targets
Accurate—identify small fold-changes with high accuracy of quantitation
Extensive content—over 1.8 million predesigned assays available for over 25 different species
Gold-standard TaqMan qPCR chemistry—TaqMan assays draw on Thermo Fisher Scientific's bioinformatics assay design pipeline to help ensure high specificity and minimal cross-reactivity, even for gene variants with high sequence homology
Ideal for multiplexing—combine one FAM-labeled and one VIC-labeled assay to create a duplex assay for two different gene targets in the same qPCR well
Ideal for control assays—use a VIC primer-limited assay for high-expressing endogenous control genes

Approximate ship time
Made to order: 4–6 days in North America, 6–10 days in Europe

Pre-formulated assay
• 2 unlabeled PCR primers (150 nM each final 1X concentration)
• 1 VIC dye-labeled TaqMan MGB probe (250 nM final 1X concentration)

TaqMan Gene Expression assays are the gold standard in real-time PCR gene expression studies, built on more than 20 years of experience. Each assay includes target primers and a sequence-specific probe optimized for the best functional performance. No additional design, optimization, or melt curve analysis is needed. Available in a wide variety of formats and species, new assay designs are constantly added to help meet your research needs. TaqMan assays have been cited in over 40,000 publications and are backed by more than 350 patents. All of our predesigned TaqMan Gene Expression assays are covered by the TaqMan Assays qPCR Guarantee.*

Recommended master mix: TaqMan Fast Advanced Master Mix

*Subject to terms and conditions. For complete details, go to www.thermofisher.com/taqmanguarantee.

For Research Use Only. Not for use in diagnostic procedures.

Specifications

Detection Method
Primer-probe
Format
Tube
GC-Rich PCR Performance
High
PCR Method
qPCR
Reaction Speed
Fast
For Use With (Equipment)
7500 System, 7500 Fast System, 7900HT System, StepOne™, StepOnePlus™, ViiA™ 7 System, QuantStudio™ 3, QuantStudio™ 5, QuantStudio™ 6 Flex, QuantStudio™ 7 Flex, QuantStudio 6 Pro, QuantStudio 7 Pro, QuantStudio™ 12k Flex
Green Features
Sustainable packaging
Product Line
TaqMan™
Product Type
Gene Expression Assay
Shipping Condition
Room Temperature
Concentration
60X
Fidelity (vs. Taq)
60 X
No. of Reactions
2900 Reactions

Contents & Storage

1 tube containing a 20X (S and M sizes) or 60X (L size) mix of pre-formulated assay (1 probe and 2 primers).

Store at -15 to -25°C.

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text