Potassium (cyanomethyl)trifluoroborate, 95%, Thermo Scientific Chemicals
Potassium (cyanomethyl)trifluoroborate, 95%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Potassium (cyanomethyl)trifluoroborate, 95%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number 444860010
Price (USD)/ Each
-
Quantity:
1 g
Request bulk or custom format
Chemical Identifiers
CAS888711-58-8
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to off-white
Appearance (Form)Solid
Infrared spectrumConforms
Melting point144°C to 156°C
NMR (1H-NMR)Consistent with structure
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only