Potassium cyclopropyltrifluoroborate, 97%, Thermo Scientific Chemicals
Potassium cyclopropyltrifluoroborate, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Potassium cyclopropyltrifluoroborate, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
Packaging:
Glass bottle
Catalog number 444870050
Price (USD)/ Each
-
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
IUPAC Namepotassium cyclopropyltrifluoroboranuide
Molecular FormulaC3H5BF3K
InChI KeyCFMLURFHOSOXRC-UHFFFAOYSA-N
SMILES[K+].F[B-](F)(F)C1CC1
Molecular Weight (g/mol)147.98
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to off-white
Appearance (Form)Solid
Infrared spectrumConforms
NMR (1H-NMR)Consistent with structure
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only