4-(Dimethylamino)phenylboronic acid pinacol ester, 97%, Thermo Scientific Chemicals
4-(Dimethylamino)phenylboronic acid pinacol ester, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

4-(Dimethylamino)phenylboronic acid pinacol ester, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
Catalog number 444960050
Price (USD)/ Each
-
Quantity:
5 g
Request bulk or custom format
Chemical Identifiers
IUPAC NameN,N-dimethyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)aniline
Molecular FormulaC14H22BNO2
InChI KeyDGMLJJIUOFKPKB-UHFFFAOYSA-N
SMILESCN(C)C1=CC=C(C=C1)B1OC(C)(C)C(C)(C)O1
Molecular Weight (g/mol)247.15
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Off-white to light green to tan
Appearance (Form)Powder or solid
Infrared spectrumConforms
Melting point122°C to 127°C
NMR (1H-NMR)Consistent with structure
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only