Thermo Scientific Chemicals

6-[3-(Dimethylamino)propoxy]pyridine-3-boronic acid pinacol ester, 97%, Thermo Scientific Chemicals

Catalog number: 444970050
5 g, Each
Thermo Scientific Chemicals

6-[3-(Dimethylamino)propoxy]pyridine-3-boronic acid pinacol ester, 97%, Thermo Scientific Chemicals

Catalog number: 444970050
5 g, Each
Quantity
Catalog number: 444970050
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
918643-56-8
IUPAC Name
dimethyl(3-{[5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridin-2-yl]oxy}propyl)amine
Molecular Formula
C16H27BN2O3
InChI Key
QWJUJXPYTLOKRL-UHFFFAOYSA-N
SMILES
CN(C)CCCOC1=NC=C(C=C1)B1OC(C)(C)C(C)(C)O1
Appearance (Color)
White to off-white
Appearance (Form)
Solid
Infrared spectrum
Conforms
Melting point
33°C to 39°C
GC
>=96.0 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text