Attune™ Focusing Fluid (1X)

Catalog number: 4449791
Have Questions?

Attune™ Focusing Fluid (1X)

Catalog number: 4449791
Have Questions?
Catalog Number
4449791
Unit Size
6 x 1 L
Price (USD)
Availability
-
Quantity
Catalog Number
Unit Size
10 L
Price (USD)
Availability
-
Quantity
Catalog Number
Unit Size
1 L
Price (USD)
Availability
-
Quantity
Catalog Number
Unit Size
20 L
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
44497916 x 1 L
-
A2490410 L
-
44886211 L
-
J10662720 L
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
Attune™ Focusing Fluid is an optimized buffer solution that is used in the Attune™ Acoustic Focusing Cytometer for focusing and transportation of particles throughout the fluidics system.

Maintains System Cleanliness
Attune™ Focusing Fluid is a sterile, buffered, azide-free support reagent that contains a surfactant that helps clean the fluidic lines of the Attune™ Acoustic Focusing Cytometer. The Attune™ Focusing Fluid was designed to minimize bubble formation and to prevent build-up of cellular material in the flow cell, resulting in less instrument clogging.

Convenient
The ready-to-use 1X focusing fluid was designed to eliminate the need for dilution prior to use, as well as access to a purified water system. The convenient 10 L packaging provides enough focusing fluid to operate for at least 10 days (the average focusing fluid consumption per day is less than 1 L) and has a spigot to to make filling of the onboard fluidics tanks easier.

High Quality
Each batch of Attune™ Focusing Fluid is traceable by its lot number designation and has been validated for use on the Attune™ Acoustic Focusing Cytometer.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

For Use With (Equipment)
Attune™ Acoustic Focusing Cytometer
Format
Liquid
Product Line
Attune™
Quantity
6 x 1 L
Product Type
Focusing Fluid

Contents & Storage

Store at 15—30°C

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text