Thermo Scientific Chemicals

1-[(Ethoxycarbonyl)methyl]-1H-pyrazole-4-boronic acid pinacol ester, 97%, Thermo Scientific Chemicals

Catalog number: 445010050
5 g, Each
Thermo Scientific Chemicals

1-[(Ethoxycarbonyl)methyl]-1H-pyrazole-4-boronic acid pinacol ester, 97%, Thermo Scientific Chemicals

Catalog number: 445010050
5 g, Each
Quantity
Catalog number: 445010050
Price (USD)
Have Questions?

Chemical Identifiers

CAS
864754-16-5
IUPAC Name
ethyl 2-[4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazol-1-yl]acetate
Molecular Formula
C13H21BN2O4
InChI Key
YUEZJHOSHBTWPV-UHFFFAOYSA-N
SMILES
CCOC(=O)CN1C=C(C=N1)B1OC(C)(C)C(C)(C)O1
Appearance (Color)
Colorless to yellow to orange
Appearance (Form)
Viscous liquid
Infrared spectrum
Conforms
GC
>=96.0 %
NMR (1H-NMR)
Consistent with structure

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text