Thermo Scientific Chemicals

2-Fluoro-5-formylphenylboronic acid pinacol ester, 95%, Thermo Scientific Chemicals

Catalog number: 445030010
1 g, Each
Thermo Scientific Chemicals

2-Fluoro-5-formylphenylboronic acid pinacol ester, 95%, Thermo Scientific Chemicals

Catalog number: 445030010
1 g, Each
Quantity
Catalog number: 445030010
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
1112208-82-8
IUPAC Name
4-fluoro-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzaldehyde
Molecular Formula
C13H16BFO3
InChI Key
WHYMLQVABFAGEK-UHFFFAOYSA-N
SMILES
CC1(C)OB(OC1(C)C)C1=C(F)C=CC(C=O)=C1
Appearance (Color)
Colorless to yellow
Appearance (Form)
Low melting solid or viscous liquid
Infrared spectrum
Conforms
GC
>=94.0 %
NMR (1H-NMR)
Consistent with structure

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text