2-Fluoro-5-formylphenylboronic acid pinacol ester, 95%, Thermo Scientific Chemicals
2-Fluoro-5-formylphenylboronic acid pinacol ester, 95%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2-Fluoro-5-formylphenylboronic acid pinacol ester, 95%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
Catalog number 445030050
Price (USD)/ Each
795.00
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
795.00
Add to cart
2-Fluoro-5-formylphenylboronic acid pinacol ester, 95%, Thermo Scientific Chemicals
Catalog number445030050
Price (USD)/ Each
795.00
-
Add to cart
Chemical Identifiers
CAS1112208-82-8
IUPAC Name4-fluoro-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzaldehyde
Molecular FormulaC13H16BFO3
InChI KeyWHYMLQVABFAGEK-UHFFFAOYSA-N
SMILESCC1(C)OB(OC1(C)C)C1=C(F)C=CC(C=O)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Colorless to yellow
Appearance (Form)Low melting solid or viscous liquid
Infrared spectrumConforms
GC>=94.0 %
NMR (1H-NMR)Consistent with structure
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only