1-Isopropyl-1H-pyrazole-4-boronic acid pinacol ester, 97%, Thermo Scientific Chemicals
1-Isopropyl-1H-pyrazole-4-boronic acid pinacol ester, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

1-Isopropyl-1H-pyrazole-4-boronic acid pinacol ester, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
Catalog number 445090050
Price (USD)/ Each
641.00
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
641.00
Add to cart
1-Isopropyl-1H-pyrazole-4-boronic acid pinacol ester, 97%, Thermo Scientific Chemicals
Catalog number445090050
Price (USD)/ Each
641.00
-
Add to cart
Chemical Identifiers
IUPAC Name1-(propan-2-yl)-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazole
Molecular FormulaC12H21BN2O2
InChI KeyOGYYMVGDKVJYSU-UHFFFAOYSA-N
SMILESCC(C)N1C=C(C=N1)B1OC(C)(C)C(C)(C)O1
Molecular Weight (g/mol)236.12
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to off-white to light brown
Appearance (Form)Solid
Infrared spectrumConforms
Melting point38°C to 46°C
GC>=96.0 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only