4-(Methylamino)phenylboronic acid pinacol ester, 97%, Thermo Scientific Chemicals
4-(Methylamino)phenylboronic acid pinacol ester, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

4-(Methylamino)phenylboronic acid pinacol ester, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number 445150010
Price (USD)/ Each
-
Quantity:
1 g
Request bulk or custom format
Chemical Identifiers
CAS845870-55-5
IUPAC NameN-methyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)aniline
Molecular FormulaC13H20BNO2
InChI KeyWWGNYCWKFZIQQS-UHFFFAOYSA-N
SMILESCNC1=CC=C(C=C1)B1OC(C)(C)C(C)(C)O1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Solid
Appearance (Color)White to brown to purple
Infrared spectrumConforms
Melting point63°C to 67°C
GC>=96 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only