Thermo Scientific Chemicals

4-(Methylamino)phenylboronic acid pinacol ester, 97%, Thermo Scientific Chemicals

Catalog number: 445150050
5 g, Each
Thermo Scientific Chemicals

4-(Methylamino)phenylboronic acid pinacol ester, 97%, Thermo Scientific Chemicals

Catalog number: 445150050
5 g, Each
Quantity
Catalog number: 445150050
Price (USD)
Have Questions?

Chemical Identifiers

CAS
845870-55-5
IUPAC Name
N-methyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)aniline
Molecular Formula
C13H20BNO2
InChI Key
WWGNYCWKFZIQQS-UHFFFAOYSA-N
SMILES
CNC1=CC=C(C=C1)B1OC(C)(C)C(C)(C)O1
Appearance (Form)
Solid
Appearance (Color)
White to brown to purple
Infrared spectrum
Conforms
Melting point
63°C to 67°C
GC
>=96 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text