2-Methyl-1H-benzimidazole-5-boronic acid pinacol ester, 97%, Thermo Scientific Chemicals
2-Methyl-1H-benzimidazole-5-boronic acid pinacol ester, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2-Methyl-1H-benzimidazole-5-boronic acid pinacol ester, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 445160010
Price (USD)/ Each
-
Quantity:
1 g
Request bulk or custom format
Chemical Identifiers
CAS1314216-34-6
IUPAC Name2-methyl-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-1,3-benzodiazole
Molecular FormulaC14H19BN2O2
InChI KeyCPQGLMCLTALRHF-UHFFFAOYSA-N
SMILESCC1=NC2=CC(=CC=C2N1)B1OC(C)(C)C(C)(C)O1
View more
SpecificationsSpecification SheetSpecification Sheet
NMR (1H-NMR)Consistent with structure
Appearance (Color)White to yellow
Appearance (Form)Solid
Infrared spectrumConforms
Melting point205°C to 211°C
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only