Thermo Scientific Chemicals

2-Methyl-1H-benzimidazole-5-boronic acid pinacol ester, 97%, Thermo Scientific Chemicals

Catalog number: 445160050
5 g, Each
Thermo Scientific Chemicals

2-Methyl-1H-benzimidazole-5-boronic acid pinacol ester, 97%, Thermo Scientific Chemicals

Catalog number: 445160050
5 g, Each
Quantity
Catalog number: 445160050
Price (USD)
Have Questions?

Chemical Identifiers

CAS
1314216-34-6
IUPAC Name
2-methyl-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-1,3-benzodiazole
Molecular Formula
C14H19BN2O2
InChI Key
CPQGLMCLTALRHF-UHFFFAOYSA-N
SMILES
CC1=NC2=CC(=CC=C2N1)B1OC(C)(C)C(C)(C)O1
NMR (1H-NMR)
Consistent with structure
Appearance (Color)
White to yellow
Appearance (Form)
Solid
Infrared spectrum
Conforms
Melting point
205°C to 211°C

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text