Thermo Scientific Chemicals

5-(Methylsulfonyl)pyridine-3-boronic acid pinacol ester, 95%, Thermo Scientific Chemicals

Catalog number: 445180010
1 g, Each
Thermo Scientific Chemicals

5-(Methylsulfonyl)pyridine-3-boronic acid pinacol ester, 95%, Thermo Scientific Chemicals

Catalog number: 445180010
1 g, Each
Quantity
Catalog number: 445180010
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
1206641-26-0
IUPAC Name
3-methanesulfonyl-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine
Molecular Formula
C12H18BNO4S
InChI Key
BFWBUXKEBXAKDE-UHFFFAOYSA-N
SMILES
CC1(C)OB(OC1(C)C)C1=CC(=CN=C1)S(C)(=O)=O
Appearance (Color)
Off-white to tan
Appearance (Form)
Solid
Melting point
164°C to 169°C
GC
>=94.0 %
NMR (1H-NMR)
Consistent with structure

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text