2-Morpholinopyridine-4-boronic acid pinacol ester, 97%, Thermo Scientific Chemicals
2-Morpholinopyridine-4-boronic acid pinacol ester, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2-Morpholinopyridine-4-boronic acid pinacol ester, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 445200050
Price (USD)/ Each
-
Quantity:
5 g
Request bulk or custom format
Chemical Identifiers
CAS888721-86-6
IUPAC Name4-[4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridin-2-yl]morpholine
Molecular FormulaC15H23BN2O3
InChI KeyHZJIGVGLNZISJU-UHFFFAOYSA-N
SMILESCC1(C)OB(OC1(C)C)C1=CC(=NC=C1)N1CCOCC1
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Appearance (Color)White to off-white to light brown
Appearance (Form)Powder or solid
GC>=96.0 %
Melting point82°C to 88°C
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only