Thermo Scientific Chemicals

2-Nitrophenylboronic acid pinacol ester, 97%, Thermo Scientific Chemicals

Catalog number: 445210050
5 g, Each
Thermo Scientific Chemicals

2-Nitrophenylboronic acid pinacol ester, 97%, Thermo Scientific Chemicals

Catalog number: 445210050
5 g, Each
Quantity
Catalog number: 445210050
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
190788-59-1
IUPAC Name
4,4,5,5-tetramethyl-2-(2-nitrophenyl)-1,3,2-dioxaborolane
Molecular Formula
C12H16BNO4
InChI Key
VLJYUDGCEKORNG-UHFFFAOYSA-N
SMILES
CC1(C)OB(OC1(C)C)C1=CC=CC=C1[N+]([O-])=O
Appearance (Color)
Light yellow to brown
Infrared spectrum
Conforms
Melting point
43°C to 50°C
GC
>=96.0 %
NMR (1H-NMR)
Consistent with structure

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text