Thermo Scientific Chemicals

Phenylboronic acid pinacol ester, 97%, Thermo Scientific Chemicals

Catalog number: 445220250
25 g, Glass bottle, Each
Thermo Scientific Chemicals

Phenylboronic acid pinacol ester, 97%, Thermo Scientific Chemicals

Catalog number: 445220250
25 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 445220250
Price (USD)
Have Questions?

Chemical Identifiers

CAS
24388-23-6
IUPAC Name
4,4,5,5-tetramethyl-2-phenyl-1,3,2-dioxaborolane
Molecular Formula
C12H17BO2
InChI Key
KKLCYBZPQDOFQK-UHFFFAOYSA-N
SMILES
CC1(C)OB(OC1(C)C)C1=CC=CC=C1
Appearance (Color)
Colorless to white
Appearance (Form)
Low melting solid or liquid
Infrared spectrum
Conforms
GC
>=96.0 %
NMR (1H-NMR)
Consistent with structure

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text