Thermo Scientific Chemicals

4-Amino-3-nitrophenylboronic acid, 97%, Thermo Scientific Chemicals

Catalog number: 445300050
5 g, Each
Thermo Scientific Chemicals

4-Amino-3-nitrophenylboronic acid, 97%, Thermo Scientific Chemicals

Catalog number: 445300050
5 g, Each
Quantity
Catalog number: 445300050
Price (USD)
Have Questions?

Chemical Identifiers

CAS
89466-07-9
IUPAC Name
(4-amino-3-nitrophenyl)boronic acid
Molecular Formula
C6H7BN2O4
InChI Key
IRTXQNNJQZNKRP-UHFFFAOYSA-N
SMILES
NC1=CC=C(C=C1[N+]([O-])=O)B(O)O
HPLC
>=96.0 %
NMR (1H-NMR)
Consistent with structure
Appearance (Color)
Orange to red
Appearance (Form)
Solid
Infrared spectrum
Conforms

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text