1x1 image pixel for data collection
5-Chloropyridine-3-boronic acid, 95%
5-Chloropyridine-3-boronic acid, 95%
5-Chloropyridine-3-boronic acid, 95%
5-Chloropyridine-3-boronic acid, 95%
Thermo Scientific Chemicals

5-Chloropyridine-3-boronic acid, 95%

CAS: 872041-85-5 | C5H5BClNO2 | 157.36 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number 445360010
Price (USD)
-
Quantity:
1 g
Request bulk or custom format
Chemical Identifiers
CAS872041-85-5
IUPAC Name(5-chloropyridin-3-yl)boronic acid
Molecular FormulaC5H5BClNO2
InChI KeyNJXYBTMCTZAUEE-UHFFFAOYSA-N
SMILESOB(O)C1=CC(Cl)=CN=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Solid
Infrared spectrumConforms
HPLC>=94.0 %
NMR (1H-NMR)Consistent with structure
Appearance (Color)White to off-white
Ask our AI about this Product
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Documents & Downloads

Certificates

Lot #Certificate TypeDateCatalog Number(s)
340909Certificate of AnalysisApr 30, 2021445360010, 445360050
1 result displayed, search above for a specific certificate

Safety Data Sheets

Share catalog number, name or link

1x1 image pixel for data collection