Thermo Scientific Chemicals

5-Chloropyridine-3-boronic acid, 95%, Thermo Scientific Chemicals

Catalog number: 445360050
5 g, Each
Thermo Scientific Chemicals

5-Chloropyridine-3-boronic acid, 95%, Thermo Scientific Chemicals

Catalog number: 445360050
5 g, Each
Quantity
Catalog number: 445360050
Price (USD)
Have Questions?

Chemical Identifiers

CAS
872041-85-5
IUPAC Name
(5-chloropyridin-3-yl)boronic acid
Molecular Formula
C5H5BClNO2
InChI Key
NJXYBTMCTZAUEE-UHFFFAOYSA-N
SMILES
OB(O)C1=CC(Cl)=CN=C1
Appearance (Form)
Solid
Infrared spectrum
Conforms
HPLC
>=94.0 %
NMR (1H-NMR)
Consistent with structure
Appearance (Color)
White to off-white

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text