The OpenArray® 384-well Sample Plates are for use with the QuantStudio™ 12K Flex Real-Time PCR System. They are low volume, barcoded microplates used in the QuantStudio™ OpenArray® Accufill™System for sample loading onto OpenArray® plates.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Barcode
Includes Barcode
Packaging
10 Plates
Product Line
OpenArray
Color
Clear
Product Type
384-Well Microplate
For Use With (Equipment)
QuantStudio™ 12k Flex
No. of Wells
384
Reaction Speed
Standard
Shipping Condition
Room Temperature
Contents & Storage
Each box contains 10 384-well plates, enough for 80 OpenArray Real-Time PCR plates.