Thermo Scientific Chemicals

4-(Methanesulfonamido)phenylboronic acid, 97%, Thermo Scientific Chemicals

Catalog number: 445470050
5 g, Each
Thermo Scientific Chemicals

4-(Methanesulfonamido)phenylboronic acid, 97%, Thermo Scientific Chemicals

Catalog number: 445470050
5 g, Each
Quantity
Catalog number: 445470050
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
380430-57-9
IUPAC Name
(4-methanesulfonamidophenyl)boronic acid
Molecular Formula
C7H10BNO4S
InChI Key
NDVJJEADFLTFCD-UHFFFAOYSA-N
SMILES
CS(=O)(=O)NC1=CC=C(C=C1)B(O)O
HPLC
>=96.0 %
Additional info
May contain varying amounts of anhydrides
Appearance (Color)
Off-white to cream to light brown
NMR (1H-NMR)
Consistent with structure
Appearance (Form)
Solid

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text